siRNA Details
| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033679.1 | NP_001028851.2 | 5, 6 | 547 | ||
| GenBank mRNA | BC091371.1 | - | 66 | |||
| BC104679.1 | - | 5, 6 | 547 | 7456971 | ||
| FQ212815.1 | - | 4, 5 | 655 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar1b| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033679.1 | NP_001028851.2 | 5, 6 | 547 | ||
| GenBank mRNA | BC091371.1 | - | 66 | |||
| BC104679.1 | - | 5, 6 | 547 | 7456971 | ||
| FQ212815.1 | - | 4, 5 | 655 | |||