siRNA Details
| Gene Symbol: Ptgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013015.2 | NP_037147.1 | 2, 3 | 284 | ||
| GenBank mRNA | FQ214359.1 | - | 1, 2 | 536 | ||
| J04488.1 | - | 2, 3 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgds| Gene Symbol: Ptgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013015.2 | NP_037147.1 | 2, 3 | 284 | ||
| GenBank mRNA | FQ214359.1 | - | 1, 2 | 536 | ||
| J04488.1 | - | 2, 3 | 284 | |||