siRNA Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138508.4 | NP_612517.2 | 11 | 1041 | ||
| GenBank mRNA | BC081713.1 | - | 11 | 1054 | 7109192 | |
| M34728.1 | - | 5 | 507 | |||
| M57453.1 | - | 11 | 1041 | |||
| M58287.1 | - | 10 | 927 | |||
| M62763.1 | - | 11 | 1068 | |||
| X60654.1 | - | 11 | 1021 | |||

Scp2
Genomic Map