siRNA Details
| Gene Symbol: Ccl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013025.2 | NP_037157.2 | 2 | 192 | ||
| GenBank mRNA | EF121994.1 | - | 2 | 123 | ||
| EF121995.1 | - | 2 | 123 | |||
| U06435.1 | - | 2 | 123 | |||
| U22414.1 | - | 2 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl3| Gene Symbol: Ccl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013025.2 | NP_037157.2 | 2 | 192 | ||
| GenBank mRNA | EF121994.1 | - | 2 | 123 | ||
| EF121995.1 | - | 2 | 123 | |||
| U06435.1 | - | 2 | 123 | |||
| U22414.1 | - | 2 | 192 | |||