siRNA Details
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138879.1 | NP_620234.1 | 5 | 815 | ||
XM_008769608.1 | XP_008767830.1 | 6 | 1197 | |||
GenBank mRNA | L25527.1 | - | 5 | 815 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138879.1 | NP_620234.1 | 5 | 815 | ||
XM_008769608.1 | XP_008767830.1 | 6 | 1197 | |||
GenBank mRNA | L25527.1 | - | 5 | 815 |