siRNA Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013031.1 | NP_037163.1 | 15 | 1481 | ||
| XM_006231664.2 | XP_006231726.1 | 15 | 2322 | |||
| XM_008760515.1 | XP_008758737.1 | 13 | 1813 | |||
| XM_008760516.1 | XP_008758738.1 | 14 | 2459 | |||
| XM_008760517.1 | XP_008758739.1 | 14 | 2454 | |||
| XM_008760518.1 | XP_008758740.1 | 13 | 1813 | |||
| XM_008760519.1 | XP_008758741.1 | 12 | 1720 | |||
| XM_008760520.1 | XP_008758742.1 | 11 | 1636 | |||
| GenBank mRNA | L00603.1 | - | 15 | 1481 | ||
| M97381.1 | - | 15 | 1452 | |||

Slc18a2
Genomic Map