siRNA Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013032.3 | NP_037164.3 | 12 | 1561 | ||
| GenBank mRNA | BC061743.1 | - | 12 | 1561 | 5599027 | |
| D63772.1 | - | 1592 | ||||
| U21104.1 | - | 12 | 1573 | |||
| U39555.1 | - | 12 | 1651 | |||
| X94255.1 | - | 12 | 1618 | |||

Slc1a1
Genomic Map