siRNA Details
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013033.2 | NP_037165.1 | 14 | 1860 | ||
| GenBank mRNA | AB000729.1 | - | 14 | 1886 | ||
| BC081827.1 | - | 14 | 1892 | 7129017 | ||
| D16101.1 | - | 14 | 1860 | |||
| U03120.1 | - | 14 | 1818 | |||

Slc5a1
Genomic Map