siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127689.1 | NP_001121161.1 | 8 | 1157 | ||
| NM_013037.1 | NP_037169.1 | 8 | 1084 | |||
| GenBank mRNA | U04317.1 | - | 8 | 1157 | ||
| U04319.1 | - | 8 | 1084 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rl1| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127689.1 | NP_001121161.1 | 8 | 1157 | ||
| NM_013037.1 | NP_037169.1 | 8 | 1084 | |||
| GenBank mRNA | U04317.1 | - | 8 | 1157 | ||
| U04319.1 | - | 8 | 1084 | |||