siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031558.3 | NP_113746.1 | 4 | 565 | ||
| GenBank mRNA | AB001349.1 | - | 521 | |||
| AB006007.1 | - | 4 | 566 | |||
| AF044081.1 | - | 505 | ||||
| AY736357.1 | - | 4 | 446 | |||
| BC088859.1 | - | 4 | 457 | 7311249 | ||
| EF091859.1 | - | 3 | 312 | |||

Star
Genomic Map