siRNA Details
| Gene Symbol: Abcc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013040.2 | NP_037172.2 | 17 | 2333 | ||
| GenBank mRNA | AB045281.1 | - | 16 | 2121 | ||
| AF019628.1 | - | 17 | 2333 | |||
| AF087838.1 | - | 17 | 2333 | |||
| D83598.1 | - | 17 | 2333 | |||

Abcc9
Genomic Map