siRNA Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017103.2 | NP_058799.2 | 2 | 171 | ||
| GenBank mRNA | BC081815.1 | - | 2 | 171 | 7110428 | |
| L46816.1 | - | 186 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tceb3| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017103.2 | NP_058799.2 | 2 | 171 | ||
| GenBank mRNA | BC081815.1 | - | 2 | 171 | 7110428 | |
| L46816.1 | - | 186 | ||||