siRNA Details
| Gene Symbol: Tff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013042.2 | NP_037174.2 | 2 | 242 | ||
| GenBank mRNA | AF012534.1 | - | 2 | 142 | ||
| BC078873.1 | - | 2 | 222 | 7127312 | ||
| BP490107.1 | - | 2 | 242 | |||
| M80826.1 | - | 2 | 221 | |||
| S49317.1 | - | 2 | 216 | |||
| U48825.1 | - | 2 | 215 | |||
| X66956.1 | - | 2 | 216 | |||

Tff3
Genomic Map