siRNA Details
| Gene Symbol: Tnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013045.1 | NP_037177.1 | 17 | 3653 | ||
| GenBank mRNA | Z18630.1 | - | 17 | 3653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnr| Gene Symbol: Tnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013045.1 | NP_037177.1 | 17 | 3653 | ||
| GenBank mRNA | Z18630.1 | - | 17 | 3653 | ||