siRNA Details
| Gene Symbol: Trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013046.3 | NP_037178.1 | 3 | 713 | ||
| XM_006236899.2 | XP_006236961.1 | 3 | 1144 | |||
| GenBank mRNA | BC161830.1 | - | 3 | 660 | 9091055 | |
| FQ220299.1 | - | 1 | 261 | |||
| FQ221570.1 | - | 3 | 689 | |||
| FQ221573.1 | - | 3 | 689 | |||
| FQ222015.1 | - | 3 | 689 | |||
| FQ222377.1 | - | 3 | 688 | |||
| FQ222984.1 | - | 3 | 687 | |||
| FQ223091.1 | - | 3 | 689 | |||
| FQ228499.1 | - | 3 | 689 | |||
| FQ228758.1 | - | 3 | 689 | |||
| FQ229082.1 | - | 3 | 689 | |||
| FQ229292.1 | - | 3 | 687 | |||
| M12138.1 | - | 3 | 680 | |||
| M36317.1 | - | 3 | 680 | |||

Trh
Genomic Map