siRNA Details
| Gene Symbol: Ttpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013048.2 | NP_037180.1 | 3 | 429 | ||
| XM_008763588.1 | XP_008761810.1 | 5 | 436 | |||
| XM_008763589.1 | XP_008761811.1 | 4 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttpa| Gene Symbol: Ttpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013048.2 | NP_037180.1 | 3 | 429 | ||
| XM_008763588.1 | XP_008761810.1 | 5 | 436 | |||
| XM_008763589.1 | XP_008761811.1 | 4 | 379 | |||