siRNA Details
| Gene Symbol: Scgb1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013051.1 | NP_037183.1 | 2 | 286 | ||
| GenBank mRNA | BC069174.1 | - | 2 | 276 | 7096714 | |
| J05536.1 | - | 2 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1a1| Gene Symbol: Scgb1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013051.1 | NP_037183.1 | 2 | 286 | ||
| GenBank mRNA | BC069174.1 | - | 2 | 276 | 7096714 | |
| J05536.1 | - | 2 | 286 | |||