siRNA Details
| Gene Symbol: Ywhaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013011.3 | NP_037143.2 | 2 | 342 | ||
| XM_006241529.2 | XP_006241591.1 | 3 | 895 | |||
| GenBank mRNA | BC070941.1 | - | 2 | 342 | 7099670 | |
| BC094305.1 | - | 2 | 647 | 7313193 | ||
| D17615.1 | - | 2 | 409 | |||
| D30740.1 | - | 2 | 322 | |||
| L07913.1 | - | 1 | 111 | |||
| U37252.1 | - | 2 | 343 | |||

Ywhaz
Genomic Map