siRNA Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013055.1 | NP_037187.1 | 15 | 2783 | ||
| XM_006242381.2 | XP_006242443.1 | 15 | 4164 | |||
| XM_006242382.2 | XP_006242444.1 | 13 | 2397 | |||
| XM_008765675.1 | XP_008763897.1 | 12 | 3127 | |||
| GenBank mRNA | AY240864.1 | - | 3 | 191 | ||
| D49785.1 | - | 15 | 2783 | |||
| FQ211695.1 | - | 15 | 2633 | |||
| FQ212412.1 | - | 13 | 3303 | |||
| FQ226670.1 | - | 3 | 325 | |||

Map3k12
Genomic Map