siRNA Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013055.1 | NP_037187.1 | 6, 7 | 1276 | ||
| XM_006242381.2 | XP_006242443.1 | 6, 7 | 2657 | |||
| XM_006242382.2 | XP_006242444.1 | 4, 5 | 890 | |||
| XM_008765675.1 | XP_008763897.1 | 3, 4 | 1620 | |||
| GenBank mRNA | D49785.1 | - | 6, 7 | 1276 | ||
| FQ211695.1 | - | 6, 7 | 1126 | |||
| FQ212412.1 | - | 5, 6 | 1629 | |||

Map3k12
Genomic Map