siRNA Details
| Gene Symbol: Id3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013058.2 | NP_037190.1 | 1 | 161 | ||
| GenBank mRNA | AF000942.1 | - | 1 | 154 | ||
| BC064658.1 | - | 1 | 161 | 5623701 | ||
| D10864.1 | - | 1 | 138 | |||
| FQ212997.1 | - | 1 | 183 | |||
| FQ216267.1 | - | 1 | 183 | |||
| FQ218468.1 | - | 1 | 181 | |||
| FQ218958.1 | - | 1 | 174 | |||
| FQ229332.1 | - | 1 | 183 | |||

Id3
Genomic Map