siRNA Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013059.1 | NP_037191.1 | 2 | 196 | ||
| GenBank mRNA | J03572.1 | - | 2 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpl| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013059.1 | NP_037191.1 | 2 | 196 | ||
| GenBank mRNA | J03572.1 | - | 2 | 196 | ||