siRNA Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013059.1 | NP_037191.1 | 6 | 766 | ||
| XM_006239136.2 | XP_006239198.1 | 6 | 1140 | |||
| GenBank mRNA | BC088399.1 | - | 6 | 789 | 7128677 | |
| J03572.1 | - | 6 | 766 | |||
| Y00714.1 | - | 5 | 616 | |||

Alpl
Genomic Map