siRNA Details
| Gene Symbol: Agrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175754.1 | NP_786930.1 | 1 | 256 | ||
| XM_008764340.1 | XP_008762562.1 | 1 | 156 | |||
| GenBank mRNA | M64780.1 | - | 1 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agrn| Gene Symbol: Agrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175754.1 | NP_786930.1 | 1 | 256 | ||
| XM_008764340.1 | XP_008762562.1 | 1 | 156 | |||
| GenBank mRNA | M64780.1 | - | 1 | 256 | ||