siRNA Details
| Gene Symbol: Fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013068.1 | NP_037200.1 | 3, 4 | 357 | ||
| GenBank mRNA | K01180.1 | - | 3, 4 | 357 | ||
| M35992.1 | - | 3, 4 | 357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp2| Gene Symbol: Fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013068.1 | NP_037200.1 | 3, 4 | 357 | ||
| GenBank mRNA | K01180.1 | - | 3, 4 | 357 | ||
| M35992.1 | - | 3, 4 | 357 | |||