siRNA Details
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038597.2 | NP_001033686.1 | 3 | 1127 | ||
NM_001038599.2 | NP_001033688.2 | 3 | 1127 | |||
NM_001038600.2 | NP_001033689.1 | 3 | 1127 | |||
NM_001038601.2 | NP_001033690.1 | 3 | 1127 | |||
NM_013071.2 | NP_037203.1 | 3 | 1127 | |||
XM_008758731.1 | XP_008756953.1 | 3 | 1273 | |||
GenBank mRNA | AY225402.1 | - | 3 | 958 | ||
AY225403.1 | - | 3 | 958 | |||
AY309000.1 | - | 3 | 958 | |||
AY309002.1 | - | 3 | 952 | |||
AY309003.1 | - | 3 | 958 | |||
AY309004.1 | - | 3 | 958 | |||
DQ680043.1 | - | 797 | ||||
EU024650.1 | - | 725 | ||||
EU024651.1 | - | 1139 | ||||
EU024652.1 | - | 1328 | ||||
EU340244.1 | - | 1147 | ||||
EU340245.1 | - | 1403 | ||||
EU340246.1 | - | 1589 | ||||
HQ699463.1 | - | 2 | 605 | |||
L13069.1 | - | 3 | 1129 | |||
L20684.1 | - | 3 | 950 | |||
L22455.1 | - | 3 | 1089 | |||
U02083.1 | - | 3 | 1130 | |||
U35424.1 | - | 3 | 989 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
