siRNA Details
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017104.1 | NP_058800.1 | 2 | 153 | ||
XM_006247265.2 | XP_006247327.2 | 2 | 222 | |||
GenBank mRNA | U37101.1 | - | 2 | 153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017104.1 | NP_058800.1 | 2 | 153 | ||
XM_006247265.2 | XP_006247327.2 | 2 | 222 | |||
GenBank mRNA | U37101.1 | - | 2 | 153 |