siRNA Details
| Gene Symbol: Otc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013078.1 | NP_037210.1 | 8 | 955 | ||
| GenBank mRNA | BC088158.1 | - | 8 | 921 | 7325875 | |
| FQ211098.1 | - | 8 | 940 | |||
| FQ219225.1 | - | 8 | 944 | |||
| FQ219657.1 | - | 8 | 918 | |||
| K00001.1 | - | 8 | 941 | |||
| K03040.1 | - | 8 | 913 | |||
| M11266.1 | - | 8 | 955 | |||
| X01976.1 | - | 8 | 941 | |||

Otc
Genomic Map