siRNA Details
| Gene Symbol: Ptprz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170685.1 | NP_001164156.1 | 12 | 2881 | ||
| NM_013080.2 | NP_037212.2 | 12 | 2881 | |||
| XM_006236137.2 | XP_006236199.1 | 12 | 2920 | |||
| GenBank mRNA | U04998.1 | - | 12 | 2881 | ||
| U09357.1 | - | 13 | 2881 | |||

Ptprz1
Genomic Map