siRNA Details
| Gene Symbol: Ptk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013081.2 | NP_037213.1 | 32 | 3545 | ||
| XM_008765546.1 | XP_008763768.1 | 36 | 3713 | |||
| GenBank mRNA | AF020777.1 | - | 32 | 3544 | ||
| CA511845.1 | - | 1 | 37 | |||
| CB713440.1 | - | 2 | 254 | |||

Ptk2
Genomic Map