siRNA Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013083.2 | NP_037215.1 | 8 | 1451 | ||
GenBank mRNA | BC062017.1 | - | 8 | 1411 | 5621611 | |
M14050.1 | - | 7 | 1327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013083.2 | NP_037215.1 | 8 | 1451 | ||
GenBank mRNA | BC062017.1 | - | 8 | 1411 | 5621611 | |
M14050.1 | - | 7 | 1327 |