siRNA Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013084.2 | NP_037216.1 | 10, 11 | 1245 | ||
| GenBank mRNA | FQ210776.1 | - | 10, 11 | 1254 | ||
| U64451.1 | - | 10, 11 | 1246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acadsb| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013084.2 | NP_037216.1 | 10, 11 | 1245 | ||
| GenBank mRNA | FQ210776.1 | - | 10, 11 | 1254 | ||
| U64451.1 | - | 10, 11 | 1246 | |||