siRNA Details
| Gene Symbol: Plau | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013085.3 | NP_037217.3 | 1001 | |||
| GenBank mRNA | BC105859.1 | - | 1001 | 7456771 | ||
| X63434.1 | - | 1046 | ||||
| X65651.1 | - | 9 | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plau| Gene Symbol: Plau | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013085.3 | NP_037217.3 | 1001 | |||
| GenBank mRNA | BC105859.1 | - | 1001 | 7456771 | ||
| X63434.1 | - | 1046 | ||||
| X65651.1 | - | 9 | 1032 | |||