siRNA Details
| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF346902.1 | - | 10 | 1268 | ||
| J05446.1 | - | 10 | 1313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gys2| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF346902.1 | - | 10 | 1268 | ||
| J05446.1 | - | 10 | 1313 | |||