siRNA Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013090.2 | NP_037222.2 | 4 | 435 | ||
| GenBank mRNA | AF498262.1 | - | 3 | 301 | ||
| BC092206.1 | - | 4 | 333 | 7313343 | ||
| CB582258.1 | - | 4 | 435 | |||
| EF653274.1 | - | 4 | 360 | |||
| EF653275.1 | - | 4 | 360 | |||
| M24104.1 | - | 4 | 398 | |||
| U74621.1 | - | 4 | 333 | |||

Vamp1
Genomic Map