siRNA Details
| Gene Symbol: Krt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199370.1 | NP_955402.1 | 5, 6 | 847 | ||
| GenBank mRNA | AY464139.1 | - | 5, 6 | 847 | ||
| BC091106.1 | - | 5, 6 | 905 | 7381703 | ||
| BC097497.1 | - | 5, 6 | 870 | 7462062 | ||
| M63482.1 | - | 5, 6 | 911 | |||
| S76054.1 | - | 5, 6 | 911 | |||

Krt8
Genomic Map