siRNA Details
| Gene Symbol: Nkx2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013093.1 | NP_037225.1 | 2 | 1979 | ||
| XM_006240116.2 | XP_006240178.1 | 3 | 2260 | |||
| XM_006240117.2 | XP_006240179.1 | 3 | 2333 | |||
| XM_006240118.2 | XP_006240180.1 | 3 | 2844 | |||
| GenBank mRNA | AB894119.1 | - | 2 | 1978 | ||
| X53858.1 | - | 2 | 2041 | |||

Nkx2-1
Genomic Map