siRNA Details
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013094.1 | NP_037226.1 | 5 | 705 | ||
GenBank mRNA | L26043.1 | - | 5 | 705 | ||
L26044.1 | - | 5 | 705 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013094.1 | NP_037226.1 | 5 | 705 | ||
GenBank mRNA | L26043.1 | - | 5 | 705 | ||
L26044.1 | - | 5 | 705 |