siRNA Details
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053297.2 | NP_445749.1 | 9 | 1366 | ||
| XM_006243187.2 | XP_006243249.1 | 9 | 1596 | |||
| XM_006243190.2 | XP_006243252.1 | 10 | 1668 | |||
| GenBank mRNA | FQ223721.1 | - | 9 | 1374 | ||
| FQ224316.1 | - | 6 | 1024 | |||
| X15800.1 | - | 9 | 1368 | |||

Pkm
Genomic Map