siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013095.3 | NP_037227.1 | 9 | 1394 | ||
| XM_008766216.1 | XP_008764438.1 | 7 | 1333 | |||
| XM_008766217.1 | XP_008764439.1 | 7 | 1059 | |||
| GenBank mRNA | BC064437.1 | - | 1394 | 5599910 | ||
| U66479.1 | - | 9 | 1203 | |||

Smad3
Genomic Map