siRNA Details
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013097.2 | NP_037229.1 | 4 | 529 | ||
XM_006245829.2 | XP_006245891.1 | 5 | 1209 | |||
GenBank mRNA | AF397150.1 | - | 4 | 529 | ||
AF397151.1 | - | 5 | 400 | |||
BC089764.1 | - | 4 | 529 | 7130586 | ||
X56060.1 | - | 4 | 382 |
