siRNA Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 13, 14 | 1760 | ||
| GenBank mRNA | AF110461.2 | - | 13, 14 | 1452 | ||
| L27057.1 | - | 13, 14 | 1760 | |||
| L27062.1 | - | 7, 8 | 1290 | |||
| L36467.1 | - | 1650 | ||||
| M25348.1 | - | 6, 7 | 720 | |||
| M26715.1 | - | 7, 8 | 1024 | |||
| M26716.1 | - | 7, 8 | 928 | |||
| M26717.1 | - | 6, 7 | 893 | |||
| M28411.1 | - | 6, 7 | 720 | |||

Pde4a
Genomic Map