siRNA Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 15 | 2396 | ||
| GenBank mRNA | AF110461.2 | - | 15 | 2088 | ||
| L27057.1 | - | 15 | 2396 | |||
| L27062.1 | - | 9 | 1926 | |||
| L36467.1 | - | 2286 | ||||
| M26715.1 | - | 9 | 1660 | |||
| M26716.1 | - | 9 | 1564 | |||
| M26717.1 | - | 8 | 1529 | |||

Pde4a
Genomic Map