siRNA Details
| Gene Symbol: Igfbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013104.3 | NP_037236.1 | 2 | 380 | ||
| GenBank mRNA | BC099742.1 | - | 2 | 392 | 7937967 | |
| FQ220221.1 | - | 2 | 450 | |||
| FQ220720.1 | - | 2 | 451 | |||
| FQ220764.1 | - | 2 | 453 | |||
| FQ230155.1 | - | 2 | 456 | |||

Igfbp6
Genomic Map