siRNA Details
| Gene Symbol: Gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013106.1 | NP_037238.1 | 4 | 536 | ||
| GenBank mRNA | DQ120473.1 | - | 4 | 429 | ||
| DQ120474.1 | - | 4 | 429 | |||
| FQ222046.1 | - | 4 | 599 | |||
| J03219.1 | - | 4 | 545 | |||
| M20713.1 | - | 4 | 536 | |||

Gnai3
Genomic Map