siRNA Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013111.3 | NP_037243.2 | 1368 | |||
| XM_008768943.1 | XP_008767165.1 | 6 | 1640 | |||
| XM_008768944.1 | XP_008767166.1 | 6 | 1640 | |||
| GenBank mRNA | AB066224.1 | - | 6 | 1125 | ||
| AF245000.1 | - | 1308 | ||||
| D67087.1 | - | 6 | 1098 | |||
| U70476.1 | - | 6 | 1177 | |||

Slc7a1
Genomic Map