siRNA Details
| Gene Symbol: Atp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013113.2 | NP_037245.2 | 3 | 442 | ||
| GenBank mRNA | BC078902.1 | - | 3 | 442 | 7111949 | |
| J02701.1 | - | 3 | 765 | |||
| M14137.1 | - | 3 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1b1| Gene Symbol: Atp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013113.2 | NP_037245.2 | 3 | 442 | ||
| GenBank mRNA | BC078902.1 | - | 3 | 442 | 7111949 | |
| J02701.1 | - | 3 | 765 | |||
| M14137.1 | - | 3 | 346 | |||