siRNA Details
| Gene Symbol: Gja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021654.2 | NP_067686.1 | 2 | 849 | ||
| GenBank mRNA | BC078837.1 | - | 2 | 849 | 7128770 | |
| BC086576.1 | - | 1 | 780 | 7306623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gja4| Gene Symbol: Gja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021654.2 | NP_067686.1 | 2 | 849 | ||
| GenBank mRNA | BC078837.1 | - | 2 | 849 | 7128770 | |
| BC086576.1 | - | 1 | 780 | 7306623 | ||