siRNA Details
| Gene Symbol: Guca2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013118.1 | NP_037250.1 | 2 | 166 | ||
| GenBank mRNA | M93005.1 | - | 2 | 166 | ||
| M95493.1 | - | 2 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Guca2a| Gene Symbol: Guca2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013118.1 | NP_037250.1 | 2 | 166 | ||
| GenBank mRNA | M93005.1 | - | 2 | 166 | ||
| M95493.1 | - | 2 | 166 | |||