siRNA Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013120.2 | NP_037252.1 | 14, 15 | 1421 | ||
| GenBank mRNA | BC081843.1 | - | 14, 15 | 1421 | 7096237 | |
| KJ026952.1 | - | 14, 15 | 1244 | |||
| X68497.1 | - | 14, 15 | 1265 | |||

Gckr
Genomic Map